Benchtop aspiration system, VACUSIP
Supplier: Integra
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
159000 159010
181-0276EA 1210 GBP
181-0276 181-0275
Benchtop aspiration system, VACUSIP
Pumps Vacuum Aspiration Systems
In the laboratory small volumes of liquid need be disposed of on a daily basis. This includes the removal of excess liquids from reaction tubes, aspiration of supernatant after centrifugation steps, removal of washing solutions from microplates and much more. This is often accomplished by self-made systems consisting of a trap flask, a rudimentary aspiration tool and an external vacuum source.
  • Small and compact; the system fits on every bench
  • Using the battery version of VACUSIP eliminates the cable mess in a laminar flow hood and increases mobility
  • The
More Product Information