VWR® Compact Vacuum Pumps For Filtration and Solid Phase Extraction
Supplier: VACUUBRAND
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
721111 20721112 721010 721011 20721012 721110
181-0239EA 0 GBP
181-0239 181-0240 181-0235 181-0236 181-0237 181-0238
VWR® Compact Vacuum Pumps For Filtration and Solid Phase Extraction
Pumps Vacuum Diaphragm Pumps
Vacuum filtration is one of the most common applications used for sample preparation in chemistry, microbiology, wastewater control and other analytical processes. VP 100 and VP 100 C diaphragm pumps offer a compact, high performance and easy to use solution which is perfect for both single and multiple filtrations.
  • Convenient, quick and simple to use due to the top-mounted power switch
  • Whisper-quiet with ultra-low vibration resulting from a patented motor mounting
  • Requires minimal bench space
  • Maintenance-free drive system and proven long diaphragm life
More Product Information