Diaphragm pumps, Dymax 30
Supplier: AUSTEN CHARLES PUMPS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
X50-001 X50-002
181-0006EA 1320 GBP
181-0006 181-0007
Diaphragm pumps, Dymax 30
Pumps Vacuum Diaphragm Pumps
The Dymax 5 provides a clean oil free vacuum or pressure source. It is rated for continuous operation and is ideal for use in education and general purpose applications such as aeration and vacuum filtration.
  • Contamination-free pumping of air and gas
  • Easy to maintain
  • Whisper quiet and ruggedly reliable
Vacuum tweezer option:
Pump as above but supplied with a hand piece and a set of suction probes, ideal for handling small delicate objects such as gold leaf. The integral filter is washable so it is also ideal for precision cleaning.