Peristaltic pumps, single-channel, P-1
Supplier: Cytiva
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
18-1110-91
18-1110-91EA 2731 GBP
18-1110-91
Peristaltic pumps, single-channel, P-1
Pumps Peristaltic Pumps
Single channel laboratory pump for use in liquid chromatography and other applications where accurately controlled liquid flows are required.
Single-channel peristaltic pump combines reproducible flow rates with simple operation, either as a standalone unit or interfaced with programmable fraction collectors. Features low pulsation flow from 0,6 to 500 ml/h, automatic pressure plate for convenient tubing changes as well as maximum and reverse flow functions.
Ordering information: Supplied with support rod/P-1, silicone tubing 1 mm, 2,1 mm and 3,1 mm I-Ø (each 2), six tubing connectors,More Product Information