Supelco® Organic Reference Standard, Lipid Standards: Triglyceride Mixtures
Supplier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
17811-1AMP
17811-1AMPEA 70.6 GBP
17811-1AMP
Supelco® Organic Reference Standard, Lipid Standards: Triglyceride Mixtures
Standards Organic Standards
Packaging: 100 mg ampule contains approximately equal amounts by weight of: Tricaprylin (C8:0) Tricaprin (C10:0) Trilaurin (C12:0) Trimyristin (C14:0) Tripalmitin (C16:0)