n-Pentane ≥98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Pentane

17381.AK 17381.K2
17381.AKEA 21.4 GBP
17381.AK 17381.K2
n-Pentane ≥98%
n-Pentane

Purity: 98% CAS No.: 109-66-0 Molecular Formula: C5H12 Formula Weight: 72.15 Flash Point: -49°(-57°F) Physical Form: Liquid Density: 0.626 Refractive Index: 1.3580 Appearance: Colourless Odour: Nearly Odourless Melting Point: -130° Boiling Point: 36° Merck Reference: 14,7116 UN No.: 1265 MDL No.: MFCD00009498

Formula: H₃C(CH₂)₃CH₃
Boiling Pt: 36 °C
Melting Pt: –130 °C
Density: 0.626
Flash Pt: –49 °C (–57 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009498
CAS Number: 109-66-0
EINECS: 203-692-4
UN: 1265
ADR: 3,II
REACH: 01-2119459286-30
Merck Index: 14,7116
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ba2c9c81a4b1f44769627a00ba49b6cc8e63132&vid=f6831eb15ab1602d8c752468dc3ee31cc4ee61c0&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR