Ultrapure water systems, arium® pro

Supplier: Sartorius
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

H2OPRO-DI-T H2OPRO-UF-B H2OPRO-UF-T H2OPRO-UV-B H2OPRO-UV-T H2OPRO-VF-B H2OPRO-VF-T H2OPRO-DI-B H2OPRO-UV-BTOC H2OPRO-UV-TTOC H2OPRO-VF-BTOC H2OPRO-VF-TTOC
171-0394EA 9130 GBP
171-0394 171-0382 171-0384 171-0385 171-0387 171-0388 171-0390 171-0648 171-0558 171-0557 171-0561 171-0560
Ultrapure water systems, arium® pro
Water Purification Systems Pure and Ultra Pure Water Systems
Arium® pro Type I water purification systems are designed for sophisticated users to meet all their requirements for reagent grade water, needed both for critical applications and routine analyses. Different versions are available for reliable and optimal analytical results.

  • Compact design, flexibility of use and simplicity of maintenance
  • Innovative glass operator interface with touch function
  • Service and alarm functions supported by graphic displays
  • Conductivity measurement for feed and product water
  • Integrated monitoring of water consumption
  • RNases: <0
More Product Information
Order Now

SPECIFICATIONS

Model arium® pro UF arium® pro DI arium® pro UV arium® pro VF
Environmentally Preferable
Flow rate Up to 1,7 l/min Up to 2 l/min Up to 2 l/min Up to 1,7 l/min
TOC content 1 - 4 ppb at 100 ppb feed water 1 - 4 ppb at 100 ppb feed water 1 - 4 ppb at 100 ppb feed water 1 - 4 ppb at 100 ppb feed water
Resistivity at +25 °C 18,2 MΩ.cm 18,2 MΩ.cm 18,2 MΩ.cm 18,2 MΩ.cm
Particles <1/ml (@ 0,2 μm) <1/ml (@ 0,2 μm) <1/ml (@ 0,2 μm) <1/ml (@ 0,2 μm)
Endotoxin <0,001 EU/ml N/A N/A <0,001 EU/ml
Microorganisms <1 CFU/1000 ml <1 CFU/1000 ml <1 CFU/1000 ml <1 CFU/1000 ml
W×D×H 350×451×492 mm 350×451×492 mm 350×451×492 mm 350×451×492 mm
Operating weight with full tank ~27 kg ~27 kg ~27 kg ~27 kg
Weight ~17 kg ~17 kg ~17 kg ~17 kg

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR