Rehydration reagent, DeStreak

Supplier: Cytiva
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

17-6003-18 17-6003-19
17-6003-18EA 66.64 GBP
17-6003-18 17-6003-19
Rehydration reagent, DeStreak
Electrophoresis Reagents Western Blotting Reagents

The appearance of streaks that distort 2-D electrophoresis maps is a common problem, occurring most frequently when running gels that contain regions greater than pH 7,0. Increased sample load, increased length of the IPG strip, or using a narrower pH gradient worsens the problem. Extra spots on 2-D gels, caused by non specific oxidation of proteins, is another difficulty encountered when running gels containing basic regions. Both streaking and non specific oxidation result in poorly resolved protein patterns and reduced reproducibility between electrophoresis runs.


  • Improves reproducibility and quality of 2-D gels by preventing streaking
  • Eliminates extra spots caused by non specific oxidation of proteins
  • Stabilises proteins throughout entire 2-D runs, ensuring simplified, well-resolved protein patterns
  • Highly suitable for both analytical and preparative gels


DeStreak™ Rehydration solution effectively eliminates non specific oxidation by maintaining protein thiol groups in a single oxidation state, irrespective of sample load, pH range, or run length. Protein patterns are stabilised in IPG strips of any length and pH gradient ensuring the same stable and reproducible pattern in every analysis. The effectiveness of DeStreak™ Rehydration solution is maintained at the high sample loads and long run times required for preparative gel electrophoresis.


DeStreak Rehydration Solution contains optimized concentrations of urea, thiourea, CHAPS, and DeStreak Reagent. The solution is ready for use after addition of the appropriate IPG buffer. DeStreak Reagent is also available separately in a 1-ml pack size.

Order Now

SPECIFICATIONS

Description DeStreak Reagent DeStreak Rehydration Solution
Environmentally Preferable
Application DIGE approved DIGE approved
Colour Colorless Blue
Physical form Liquid Liquid
Shelf life One Year One Year
Storage conditions 2 - 8°C -20°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR