Ion exchange chromatography media, DEAE Sephacel™
Supplier: Cytiva
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
17-0500-01
17-0500-01EA 408 GBP
17-0500-01
Ion exchange chromatography media, DEAE Sephacel™
Protein Purification Protein Purification Resins
DEAE Sephacel™ is a weak anion exchanger based on beaded cellulose. The ion exchange group is diethylaminoethyl, which remains charged and maintains consistently high capacity over the entire working range. This media is macroporous and has an exclusion limit of ~10000 kDa for globular proteins. Supplied pre-swollen in 20% ethanol.
The ion exchange group is diethylaminoethyl, which remains charged and maintains consistently high capacity over the entire working range. This media is macroporous and has an exclusion limit of approximately 1000000 daltons for globular proteins. Supplied pre-swollen in 20% ethanol. Autoclavable in 0.1 M NaCl at 121 °C for 30 min. Avoid using oxidizing materials. Stable in aqueous buffers, 1.0 M acetic acid, 1.0 M NaOH, 8 M Urea, 8 M guanidine hydrochloride, ethanol, methanol. Negligible volume variation due to changes in pH or ionic strength.