Sepharose™ 6B Gel Filtration Base Matrix
Supplier: Cytiva
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
17-0110-01
17-0110-01EA 815 GBP
17-0110-01
Sepharose™ 6B Gel Filtration Base Matrix
Protein Purification Protein Purification Resins
Sepharose™ 6B is an agarose gel filtration base matrix and is frequently used for coupling affinity ligands to the matrix. The matrix is not pre-activated and the user performs all steps in coupling.
  • 6% agarose gel filtration media
  • Proven base matrix for coupling affinity ligands