Diethyl ether, anhydrous ≥99% stabilised ACS

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger


Stabilisation: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

16767.18 16767.36 16767.62
16767.18EA 32.6 GBP
16767.18 16767.36 16767.62
Diethyl ether, anhydrous ≥99% stabilised ACS
Diethyl ether

Purity: 99% min CAS No.: 60-29-7 Molecular Formula: C4H10O Formula Weight: 74.12 Flash Point: -45°(-49°F) Physical Form: Liquid Density: 0.713 Refractive Index: 1.3525 Appearance: Colourless Odour: Sweet Melting Point: -116° Boiling Point: 34.6° Merck Reference: 14,3806 UN No.: 1155 MDL No.: MFCD00011646

Formula: (CH₃CH₂)₂O
Boiling Pt: 34.6 °C
Melting Pt: –116 °C
Density: 0.713
Flash Pt: –45 °C (–49 °F)
Storage Temperature: Ambient
MDL Number: MFCD00011646
CAS Number: 60-29-7
EINECS: 200-467-2
UN: 1155
ADR: 3,I
REACH: 01-2119535785-29
Merck Index: 14,3806
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=16f2debd1f18f9bff9952c4a6e5d599a67f94e61&vid=3dd6c545e58e36e1004e828026523fb0e3bcbbcd&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR