Tetrabutylammonium iodide 98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Tetra-n-butylammonium iodide, TBAI

161270250 161271000 161275000 161270025
161270250.EA 16 GBP
161270250. 161271000. 161275000. 161270025.
Tetrabutylammonium iodide 98%
Tetrabutylammonium iodide
Formula: C₁₆H₃₆IN
MW: 369.37 g/mol
Melting Pt: 141…150 °C
Storage Temperature: Ambient
MDL Number: MFCD00011636
CAS Number: 311-28-4
EINECS: 206-220-5

Order Now

Specification Test Results

Appearance White to cream Crystalline powder
Infrared spectrum Conforms
Melting point 141 to 150 °C
Titration Argentometric ≥97.5 %
Loss on drying ≤1 %
Residue after ignition ≤1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR