Pindolol, Reference Standard

Supplier: USP
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

1539701
1539701.EA 368 GBP
1539701.
Pindolol, Reference Standard
Standards Organic Standards

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.


Certifications: US Pharmacopea Reference Standard

Order Now

SPECIFICATIONS

Description Pindolol
Environmentally Preferable
CAS No. 13523-86-9
UN No. 2811

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR