Bis(3-methyl-1-phenyl-5-pyrazolone) ≥98.0% derivatisation reagent, for TLC, for cyanide determination, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 3,3-Dimethyl-1,1-diphenyl-1H,1H-[4,4-bipyrazole]-5,5(4H,4H)-dione, 3,3'-Dimethyl-1,1'-diphenyl-4,4'-bi(5-pyrazolone), Bispyrazolone

M-Clarity™ quality level = MQ100

15156-5G
15156-5GEA 55.5 GBP
15156-5G
Bis(3-methyl-1-phenyl-5-pyrazolone) ≥98.0% derivatisation reagent, for TLC, for cyanide determination, Supelco®
Bis(3-methyl-1-phenyl-5-pyrazolone)

It was used for spectrophotometric determination and quantization of urea in urine samples.


For determination of ammonia, or assay of cyanogenic compounds.

Formula: C₂₀H₁₈N₄O₂
MDL Number: MFCD00020765
CAS Number: 7477-67-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9c84efa3c9ccdfe13dc3995ae3a82aef974621d7&vid=dd630011f4a1c3c19fed7a06c790600984a77eca&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR