Continuous static control 724 workstation monitor
Supplier: DESCO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
724MO
151-0320EA 203 GBP
151-0320
Continuous static control 724 workstation monitor
Static Control Monitors
The SCS 724 workstation monitor continuously monitors the path-to-ground integrity of two operators and one conductive or dissipative worksurface.
  • Adjustable operator test voltage and upper limit
  • Adjustable audible alarm
  • Simple installation and setup
It has been designed and tested for use with SCS dual-wire wrist straps and worksurface mats.
Audible and visual alarms provide instant notification of static control product failures. Monitor allows for the selection of test voltages (9 or 16 V) and resistance limits (10 or 35 megohms).