Gas detection tubes for chlorine
Supplier: Dräger
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
CH24301 CH20701
151-0195EA 115 GBP
151-0195 151-0191
Gas detection tubes for chlorine
Gas Tubes
To measure the 'spot concentrations' of chlorine for use with an Accuro® pump.
Uses include determination and measurement of concentration peaks, personal exposure in the inhalation area, detection of leaks in pipelines or gas installations, air analysis in sewers, shafts and confined spaces.
Cross sensitivity: Bromine is indicated with the same sensitivity. Chlorine dioxide and nitrogen dioxide are indicated as well, but with different sensitivities.
One stroke is equivalent to a volume of 100 ±5 ml of gas passing through the tube.
The standard measuring range is calibrated at 20More Product Information