Test sets
Supplier: Dräger
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
6400090 8101735 8101736 8101770 8103170 8103380 8103410
151-0116EA 89.5 GBP
151-0116 151-0176 151-0177 151-0179 DRAE8103170 DRAE8103380 DRAE8103410
Test sets
Gas Tubes
These test sets allow for the semi quantitative measurements of five different gases at the same time.