Florisil® 30 - 60 mesh for chromatography

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

150243CEA 166 GBP
150243C
Florisil® 30 - 60 mesh for chromatography
Magnesium silicate
Formula: MgO₃Si
MDL Number: MFCD00078249
CAS Number: 1343-88-0
EINECS: 215-681-1

Order Now

Specification Test Results

Identification Passes test
Particle size (30-60 US Mesh) Min. 70 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR