Silicon nitride α-phase ≥99.9% (metals basis)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Silicon(IV) nitride

14704.14 14704.22 14704.36
14704.14EA 43.4 GBP
14704.14 14704.22 14704.36
Silicon nitride α-phase ≥99.9% (metals basis)
Silicon nitride

Purity: 99.9% CAS No.: 12033-89-5 Molecular Formula: Si3N4 90% min (alpha form) Formula Weight: 140.28 Physical Form: 1 Micron Powder Density: 3.2 Appearance: Pale gray Odour: Odourless Melting Point: 1900° subl. Merck Reference: 14,8497 MDL No.: MFCD00011230

Formula: N₄Si₃
Melting Pt: 1900 °C (subl.)
Density: 3.2
Storage Temperature: Ambient
MDL Number: MFCD00011230
CAS Number: 12033-89-5
EINECS: 234-796-8
Merck Index: 14,8497

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR