Sodium nitrate ≥99.0% ACS

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

14493.14 14493.30 14493.A1 14493.A7
14493.14EA 24.8 GBP
14493.14 14493.30 14493.A1 14493.A7
Sodium nitrate ≥99.0% ACS
Sodium nitrate

In manufacturing of HNO₃, as a catalyst to manufacture H₂SO₄, in manufacturing of glass, enamel for pottery, sodium nitrite


Purity: 99.0% min CAS No.: 7631-99-4 Molecular Formula: NaNO3 Formula Weight: 84.99 Physical Form: Crystalline Density: 2.26 Appearance: White Odour: Odourless Melting Point: 308° Solubility: Soluble in water. Slightly soluble in methanol Merck Reference: 14,8647 UN No.: 1498 MDL No.: MFCD00011119


In manufacturing of HNO3, as a catalyst to manufacture H2SO4, in manufacturing of glass, enamel for pottery, sodium nitrite

Formula: NaNO₃
Boiling Pt: 380 °C (1013 hPa)
Melting Pt: 308 °C
Density: 2.26
Storage Temperature: Ambient
MDL Number: MFCD00011119
CAS Number: 7631-99-4
EINECS: 231-554-3
UN: 1498
ADR: 5.1,III
REACH: 01-2119488221-41
Merck Index: 14,8647

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR