Tin(II) iodide, ultra dry ≥99.999% (metals basis)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Tin(II) iodide, Tin diiodide

Ampuled under argon

14476.06 14476.14
14476.06EA 167 GBP
14476.06 14476.14
Tin(II) iodide, ultra dry ≥99.999% (metals basis)
Tin (II) iodide

Purity: 99.999% CAS No.: 10294-70-9 Molecular Formula: SnI2 Formula Weight: 372.50 Physical Form: -10 Mesh Beads, Ampouled under argon Density: 5.28 Melting Point: 320° Boiling Point: 720° dec. Merck Reference: 14,8785 UN No.: 3260 MDL No.: MFCD00049545

Formula: I₂Sn
MW: 372.52 g/mol
Boiling Pt: 720 °C (dec.)
Melting Pt: 320
Density: 5.28
Storage Temperature: Ambient
MDL Number: MFCD00049545
CAS Number: 10294-70-9
EINECS: 233-667-3
UN: 3260
ADR: 8 III
Merck Index: 14,8785

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR