Manganese(II) chloride, ultra dry ≥99.99% (metals basis)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Manganese dichloride

ampouled under argon

14464.06 14464.14 14464.22
14464.06EA 62.2 GBP
14464.06 14464.14 14464.22
Manganese(II) chloride, ultra dry ≥99.99% (metals basis)
Manganese (II) chloride

Purity: 99.99% CAS No.: 7773-01-5 Molecular Formula: Cl2Mn Formula Weight: 125.84 Physical Form: -10 Mesh Beads, Ampouled under argon Density: 2.977 Appearance: Pink Odour: Odourless Melting Point: 650° Boiling Point: 1190° Merck Reference: 14,5728 UN No.: 3288 MDL No.: MFCD00011114

Formula: MnCl₂
Boiling Pt: 1190 °C
Melting Pt: 650 °C
Density: 2.977
Storage Temperature: Ambient
MDL Number: MFCD00011114
CAS Number: 7773-01-5
EINECS: 231-869-6
UN: 3288
ADR: 6.1,III
Merck Index: 14,5728

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR