Calcium ≥99.98% (metals basis), crystalline dendritic pieces

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger


ampouled under argon

14457.06 14457.14
14457.06EA 189 GBP
14457.06 14457.14
Calcium ≥99.98% (metals basis), crystalline dendritic pieces
Calcium

Purity: 99.98% CAS No.: 7440-70-2 Physical Form: Ampouled under argon Odour: Odourless UN No.: 1401 MDL No.: MFCD00085314

Formula: Ca
Boiling Pt: 1484 °C (1013 hPa)
Melting Pt: 850 °C
Density: 1.54 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00085314
CAS Number: 7440-70-2
EINECS: 213-179-5
UN: 1401
ADR: 4.3,II
Merck Index: 13,01644

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR