Sulphur standard solution, 10000 mg/L S in water (from (NH₄)₂SO₄), standard for ICP, Specpure®

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

14436.AD 14436.AE 14436.AP
14436.ADEA 60.3 GBP
14436.AD 14436.AE 14436.AP
Sulphur standard solution, 10000 mg/L S in water (from (NH₄)₂SO₄), standard for ICP, Specpure®
Standards Cationic Standards Sulphur Standards

Molecular Formula: (NH₄)₂SO₄ in H₂O
Physical Form: Liquid
MDL No.: MFCD00085316


Certifications: Traceable to NIST

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR