Chromium standard solution, 10000 mg/L Cr in 5% hydrochloric acid (from Cr), standard for ICP, Specpure®

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

14408.AD 14408.AE 14408.AP
14408.ADEA 69 GBP
14408.AD 14408.AE 14408.AP
Chromium standard solution, 10000 mg/L Cr in 5% hydrochloric acid (from Cr), standard for ICP, Specpure®
Standards Cationic Standards Chromium Standards

Molecular Formula: Cr in 5% HCl
Physical Form: Liquid
UN No.: 1789
MDL No.: MFCD00010944


Certifications: Traceable to NIST

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR