Zinc standard solution, 10000 mg/L Zn in 5% nitric acid (from Zn), standard for ICP, Specpure®

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

14404.AD 14404.AE 14404.AP
14404.ADEA 65.6 GBP
14404.AD 14404.AE 14404.AP
Zinc standard solution, 10000 mg/L Zn in 5% nitric acid (from Zn), standard for ICP, Specpure®
Standards Cationic Standards Zinc Standards

Molecular Formula: Zn in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011291


Certifications: Traceable to NIST

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR