Platinum standard solution, 10000 mg/L Pt in 20% hydrochloric acid (from Pt), standard for ICP, Specpure®

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

14397.AD 14397.AE 14397.AP
14397.ADEA 240 GBP
14397.AD 14397.AE 14397.AP
Platinum standard solution, 10000 mg/L Pt in 20% hydrochloric acid (from Pt), standard for ICP, Specpure®
Standards Cationic Standards Platinum Standards

Molecular Formula: Pt in 20% HCl
Physical Form: Liquid
Odour: Characteristic
UN No.: 1789
MDL No.: MFCD00011179


Certifications: Traceable to NIST

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR