Terbium standard solution, 10000 mg/L Tb in 5% nitric acid (from Tb₄O₇), standard for ICP, Specpure®

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

14383.AD 14383.AE 14383.AP
14383.ADEA 76.6 GBP
14383.AD 14383.AE 14383.AP
Terbium standard solution, 10000 mg/L Tb in 5% nitric acid (from Tb₄O₇), standard for ICP, Specpure®
Standards Cationic Standards Terbium Standards

Molecular formula: Tb₄O₇ in 5% HNO₃
Physical form: liquid
UN No.: 3264
MDL No.: MFCD00011256


Traceable to NIST


Certifications: Traceable to NIST.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR