Strontium standard solution, 10000 mg/L Sr in 5% nitric acid (from Sr(NO₃)₂), standard for ICP, Specpure®

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

14382.AD 14382.AE 14382.AP
14382.ADEA 70.2 GBP
14382.AD 14382.AE 14382.AP
Strontium standard solution, 10000 mg/L Sr in 5% nitric acid (from Sr(NO₃)₂), standard for ICP, Specpure®
Standards Cationic Standards Strontium Standards

Molecular Formula: Sr(NO₃)₂ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134060


Certifications: Traceable to NIST

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR