Cadmium(II) telluride ≥99.999% (metals basis)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

14365.06 14365.14 14365.22
14365.06EA 59.3 GBP
14365.06 14365.14 14365.22
Cadmium(II) telluride ≥99.999% (metals basis)
Cadmium (II) telluride

Purity: 99.999% CAS No.: 1306-25-8 Molecular Formula: CdTe Formula Weight: 240.00 Physical Form: Polycrystalline Lump Density: 6.20 Appearance: Black Odour: Odourless Melting Point: 1041° Merck Reference: 14,1629 UN No.: 3077 MDL No.: MFCD00015998

Formula: CdTe
Melting Pt: 1041 °C
Density: 6.20
Storage Temperature: Ambient
MDL Number: MFCD00015998
CAS Number: 1306-25-8
EINECS: 215-149-9
UN: 3077
ADR: 9,III
Merck Index: 14,1629

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR