Gold ≥99.9% (metals basis), wire, Length: 4.88m (16ft), Ø 0.0127 mm (0.0005 in)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

14353.KS
14353.KSEA 200 GBP
14353.KS
Gold ≥99.9% (metals basis), wire, Length: 4.88m (16ft), Ø 0.0127 mm (0.0005 in)
Gold

Purity: 99.9% CAS No.: 7440-57-5 Physical Form: ≈0.0024g/m Odour: Odourless MDL No.: MFCD00003436

Formula: Au
Boiling Pt: 2800 °C (1013 hPa)
Melting Pt: 1063 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR