Barium standard dissolved in oil, 1,000 µg/g Ba in mineral oil for wear metal analysis, Specpure®

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

14230.18
14230.18EA 56.7 GBP
14230.18
Barium standard dissolved in oil, 1,000 µg/g Ba in mineral oil for wear metal analysis, Specpure®
Standards Cationic Standards Barium Standards

Physical Form: Liquid
MDL No.: MFCD00134031

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR