Silicon carbide, β-phase ≥99.8% (metals basis)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

14165.22 14165.36 14165.A3
14165.22EA 73.5 GBP
14165.22 14165.36 14165.A3
Silicon carbide, β-phase ≥99.8% (metals basis)
Silicon carbide, β-phase

Purity: 99.8% CAS No.: 409-21-2 Molecular Formula: SiC Formula Weight: 40.10 Physical Form: 1 Micron Powder, S.A. 14-19m2/g Odour: Odourless Merck Reference: 14,8492 MDL No.: MFCD00049531

Formula: SiC
Melting Pt: 2700 °C
Storage Temperature: Ambient
MDL Number: MFCD00049531
CAS Number: 409-21-2
EINECS: 206-991-8
Merck Index: 14,8492

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR