Sodium carbonate monohydrate 85

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Sodium carbonate hydrous, Soda monohydrate

14108.A1 14108.A7
14108.A1EA 28.9 GBP
14108.A1 14108.A7
Sodium carbonate monohydrate 85
Sodium carbonate monohydrate

Purity: 85% min CAS No.: 5968-11-6 Molecular Formula: Na2CO3.H2O Formula Weight: 124.01 (105.99anhy) Physical Form: Crystalline Density: 2.250 Appearance: Colourless to white Odour: Odourless Melting Point: 100° -H2O Solubility: Soluble in water. Insoluble in alcohol Merck Reference: 14,8596 MDL No.: MFCD00149177

Formula: Na₂CO₃·1H₂O
MW: 124.00 g/mol
Melting Pt: 100 °C (-H₂O)
Density: 2.250
Storage Temperature: Ambient
MDL Number: MFCD00149177
CAS Number: 5968-11-6
EINECS: 207-838-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR