MoonBeam™3 Disinfection Technology
Supplier: Diversey
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
DMOONBEAM3GB
141-1546EA 0 GBP
141-1546
MoonBeam™3 Disinfection Technology
Disinfection Systems
MoonBeam™3 Disinfection Technology is a UV-C device that provides an additional reduction of contamination on pre-cleaned and disinfected medical devices and surfaces, to give additional assurances for various facilities. It is designed for fast, terminal disinfection of 'high touch' surfaces in classrooms, offices, patient rooms, operating rooms and bathrooms, and can be used on equipment, fixtures, keyboards, monitors and workstations.
  • Highly effective – UV-C light provides optimal terminal disinfection in just 3 minutes
  • Portable unit can move around
More Product Information