Disinfectants, ClearKlens Cleansinald, sterile
Supplier: Diversey
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
100848254 100848253 7516429
DIVY100848254EA 369 GBP
DIVY100848254 DIVY100848253 141-0893
Disinfectants, ClearKlens Cleansinald, sterile
Cleaning Agents Disinfectants
Surface active disinfectant based on a blend of quaternary ammonium and alkyl amines.
  • 0,2 µm filtered, aseptically filled and double bagged
  • Broad spectrum of activity
  • Supplied with certificate of analysis and sterility where applicable
  • Compatible with ClearKlens Tego® 2000, ClearKlens Tego® 2001 and ClearKlens Bi-Spore as a rotational process
Certifications: Passes CEN 1276, CEN 1650 and CEN 13697
Caution: Use biocides safely. Always read the label and product information before use.
Articles may not be available in all countries. Please contactMore Product Information