Lithium fluoride ≥99.5%, Optical Grade Optical Grade

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

14056.06 14056.22
14056.06EA 30.1 GBP
14056.06 14056.22
Lithium fluoride ≥99.5%, Optical Grade Optical Grade
Lithium fluoride

Purity: 99.5+% CAS No.: 7789-24-4 Molecular Formula: LiF Formula Weight: 25.94 Physical Form: Broken Crystals Density: 2.635 Refractive Index: 1.3915 Odour: Odourless Melting Point: 845° Boiling Point: 1680° Merck Reference: 14,5531 UN No.: 3288 MDL No.: MFCD00011090

Formula: FLi
Boiling Pt: 1680 °C
Melting Pt: 845 °C
Density: 2.635
Storage Temperature: Ambient
MDL Number: MFCD00011090
CAS Number: 7789-24-4
EINECS: 232-152-0
UN: 3288
ADR: 6.1,III
Merck Index: 14,5531

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR