Transport carriers, Nalgene®
Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
7130-0010 7134-0010
139-3110EA 33.8 GBP
139-3110 215-8118
Transport carriers, Nalgene®
Carriers Transport Carriers
These bottle carriers allow for safe transport and storage of equipment, such as bottles, and containers. They include a partition wall and an integral carrying handle.
  • Good chemical resistance
  • Heat resistant up to +120 °C
  • Rigid, with two liquid tight compartments
Colour: white.