Multiple socket with overvoltage protection, 6 places, Professional, Bachmann
Supplier: BACHMANN ELEKTRO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
331.013 331.014
139-1387EA 109 GBP
139-1387 139-1392
Multiple socket with overvoltage protection, 6 places, Professional, Bachmann
Electrical Sockets
As for Professional, but also with:
  • High-performance surge protection with three varistors
  • Optical failure indicator