Shelving systems, solid, Super Erecta®
Supplier: Metro International
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1842FS 1848NFS 1860NFS 1442FG 1824FS 1430FS 1448FS 2136NFS 2142FG 2142FS 2142LS 2142NFS 2148FG 2148FS 2136LS 2148NFS 2160FG 2160FS 2160LS 2160NFS 2148LS 1848FS 1824FG 1824LS 1824NFS 1836FG 1836FS 1836LS 1836NFS 2136FS 1848FG 1848LS 1860FG 1860FS 1860LS 2436LS 2136FG 2436FG 2436NFS 2442FG 2442FS 2442LS 2442NFS 2448FG 2448FS 2448NFS 2436FS 2460FG 2460FS 2460LS 2460NFS 2448LS 1842NFS 1842FG 1842LS 1460FS 1436FG 1830FG 1830FS 1424FS 1442FS 1436FS
139-0319EA 527 GBP
139-0319 139-0356 139-0357 METO1442FG METO1824FS 132-1424 METO1448FS 139-0358 139-0334 139-0322 139-0346 139-0359 139-0335 139-0323 139-0345 139-0360 139-0336 139-0324 139-0348 139-0361 139-0347 139-0320 METO1824FG METO1824LS METO1824NFS 139-0329 139-0318 139-0341 139-0354 139-0321 139-0331 139-0343 139-0332 METO1860FS 139-0344 139-0349 139-0333 139-0337 139-0362 139-0338 139-0326 139-0351 139-0363 139-0339 139-0327 139-0364 139-0325 139-0340 139-0328 139-0353 139-0365 139-0352 139-0355 139-0330 139-0342 METO1460FS METO1436FG METO1830FG METO1830FS METO1424FS METO1442FS METO1436FS
Shelving systems, solid, Super Erecta®
Furniture Shelves
Perfect for applications involving spillage. Provides barrier between floor and bottom shelf contents to deter contamination.
  • Shelf design features a 3,2 mm raised 'ship’s edge' on all four sides to contain spills
  • Louvered/embossed shelf allows air circulation
  • Available in several styles and materials for a variety of applications
  • Shelves are constructed with 18-gauge stainless steel (type 304) or galvanized materials
Standard stainless steel solid shelves (Type 304): with epoxy-coated cast corners address the majority of applications for solid,More Product Information