Self-retracting lifelines, Miller® Falcon™
Supplier: HONEYWELL SAFETY
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1011746 1011742 1011728 1011750 1011751 1011743
113-1268EA 1730 GBP
113-1268 139-0230 139-0229 139-0231 111-1404 111-1403
Self-retracting lifelines, Miller® Falcon™
Fall Protection Fall Arresters
These self-retracting lifelines are the ideal fall protection device for use at low heights, where lanyards with shock absorbers cannot generally be used. The self-retracting lifelines can be used in many different contexts, e.g. the construction industry, manufacturing, supply and disposal.
  • Virtually indestructible, impact resistant nylon housing
  • Rapid response brake system halts a fall within only a few centimetres
  • Hub made from heat treated aluminium
  • Mechanical parts: bronze and rust-free steel
Certifications: EN 360