Chairs and Stools, Wood
Supplier: Kango
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
5NN10GHJR00000 5NN10GHJR02000 5NN35GDLV00000 5NN35GHLP00000 5NN35GBLR00000 5NN35GHLR02000 5NN35GHLV02000 5NN35NBPR00000 5NN35NBPP00000 5NN10NHNR02000 5NN10NHNR00000 5NN10NHNP02000 5NN10NHNP00000 5NN10NBNP00000 5NN10NBNR00000 5NN35GTLP01000 5NN10GBJP00000 5NN35GHLP02000 5NN35GDLV02000 5NN10GHJP02000 5NN10GBJR00000 5NN10GDJP02000 5NN35NHPR02000 5NN35NHPP02000 5NN35NHPP00000
132-7853EA 200 GBP
132-7853 132-7856 132-8092 132-8409 132-8411 BIEN5NN35GHLR02000 BIEN5NN35GHLV02000 139-0106 BIEN5NN35NBPP00000 139-0712 139-0711 139-0710 139-0709 139-0708 139-0104 BIEN5NN35GTLP01000 139-0098 BIEN5NN35GHLP02000 BIEN5NN35GDLV02000 139-0707 139-0099 139-0706 BIEN5NN35NHPR02000 139-0206 BIEN5NN35NHPP00000
Chairs and Stools, Wood
Chairs
Chairs and stools with multi-ply varnished beech wood seats/backrests. Ergonomically designed seat and backrest. Double curved seat with anti-slip varnish.
  • Ecological
  • Easily adjustable
  • Dust resistant
  • Gas lift column with height adjustment and shock absorber
  • 5-branch scuff resistant reinforced base
Chairs:
Shaped backrest and seat with rounded edges
Articulated backrest, adjustable in height and depth
Rectangular seat with double curve for ergonomics and comfort
Made of multiply beech
Moisture resistant colourless polyurethane varnish
Dimensions of the seatMore Product Information