Strontium standard solution, 1000 mg/L Sr in 5% nitric acid (from Sr(NO₃)₂), standard for ICP, Specpure®

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

13874.AD 13874.AE 13874.AP
13874.ADEA 42.7 GBP
13874.AD 13874.AE 13874.AP
Strontium standard solution, 1000 mg/L Sr in 5% nitric acid (from Sr(NO₃)₂), standard for ICP, Specpure®
Standards Cationic Standards Strontium Standards

Molecular Formula: Sr(NO₃)₂) in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134060


Certifications: Traceable to NIST

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR