Calcium standard solution, 1000 mg/L Ca in 5% nitric acid (from CaCO₃), standard for ICP, Specpure®

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

13852.AD 13852.AE 13852.AP
13852.ADEA 40 GBP
13852.AD 13852.AE 13852.AP
Calcium standard solution, 1000 mg/L Ca in 5% nitric acid (from CaCO₃), standard for ICP, Specpure®
Standards Cationic Standards Calcium Standards

Molecular Formula: CaCO₃ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00085314


Certifications: Traceable to NIST

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR