Silver standard solution, 1000 mg/L Ag in 5% nitric acid (from Ag), standard for ICP, Specpure®

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

13849.AD 13849.AE 13849.AP
13849.ADEA 45.1 GBP
13849.AD 13849.AE 13849.AP
Silver standard solution, 1000 mg/L Ag in 5% nitric acid (from Ag), standard for ICP, Specpure®
Standards Cationic Standards Silver Standards

Molecular Formula: Ag in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00003397


Certifications: Traceable to NIST

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR