Indium standard solution, 1000 mg/L In in 5% nitric acid (from In), standard for ICP, Specpure®

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

13846.AD 13846.AE 13846.AP
13846.ADEA 50.6 GBP
13846.AD 13846.AE 13846.AP
Indium standard solution, 1000 mg/L In in 5% nitric acid (from In), standard for ICP, Specpure®
Standards Cationic Standards Indium Standards

Molecular Formula: In in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134048


Certifications: Traceable to NIST

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR