Potassium hydrogen sulphate mixture of potassium pyrosulphate, K2S2O7, and potassium bisulphate, KHSO4 ACS, fused

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

13800.22 13800.36
13800.22EA 49.1 GBP
13800.22 13800.36
Potassium hydrogen sulphate mixture of potassium pyrosulphate, K2S2O7, and potassium bisulphate, KHSO4 ACS, fused
Potassium hydrogen sulphate

Formula Weight: 136.17 Physical Form: Fused Powder Sensitivity: Hygroscopic Appearance: White Odour: Odourless Melting Point: 214° UN No.: 2509 MDL No.: MFCD00066385

Formula: KHSO₄
Melting Pt: 214 °C
Density: 2.32 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011404
CAS Number: 7646-93-7
EINECS: 231-594-1
UN: 2509
ADR: 8,II
Merck Index: 13,07695

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR