Supelpak™-2SV, Pore size: 90 Å (mean pore size) 20-60 mesh, Supelco®
Supplier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
MQ100
13673-U 13674-U 13682-U
13673-UEA 499 GBP
13673-U 13674-U 13682-U
Supelpak™-2SV, Pore size: 90 Å (mean pore size) 20-60 mesh, Supelco®
Supelpak™-2SV
Supelpak™-2SV is purified Amberlite® XAD2 that has been specially cleaned and tested for optimal performance in the capturing and extraction of semivolatile organics. Packaged in glass containers.
  • Matrix: Styrene-divinylbenzene
  • Pore size: ~0.65 mL/g pore volume
  • surface area: ~300 m2/g