Scandium(III) carbonate hydrate ≥99.99% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

13646.03 13646.06
13646.03EA 103 GBP
13646.03 13646.06
Scandium(III) carbonate hydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Scandium(III) carbonate hydrate

Purity: 99.99% Molecular Formula: Sc2(CO3)3.xH2O Formula Weight: 269.94(anhy) Physical Form: Crystalline Melting Point: 500° dec. MDL No.: MFCD00011501

Formula: Sc₂(CO₃)₃·xH₂O
Melting Pt: >500 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011501
CAS Number: 5809-49-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR