Biological safety cabinets, class II, Herasafe™ 2025
Supplier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
51033311 51033312 51033313 51033314 51033315 51033316 51033317 51033318
135-2786EA 12110 GBP
135-2786 135-2787 135-2788 135-2789 135-2790 135-2791 135-2792 135-2793
Biological safety cabinets, class II, Herasafe™ 2025
Fume Hoods and Enclosures Biological Safety Cabinets
Maximise sample protection and user safety with an emphasis on containment, comfort and convenience.
  • SmartFlow Plus technology automatically balances performance of the dual-DC blower motors to maintain appropriate airflow rates whilst compensating for the loading condition of the protective HEPA filters
  • Unique DAVe Plus (Digital Airflow Verification) system validates protection by monitoring airflow velocities with independent pressure sensors that detect and alert to any changes across the exhaust and downflow plenums
  • Simple to use and easily
More Product Information